missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (2C8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-445H
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Immunohistochemistry, Sandwich ELISA
Specifications
| TNF-alpha | |
| Monoclonal | |
| HRP | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with Human recombinant tumor necrosis factor of alpha type. | |
| 0.2 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Sandwich ELISA | |
| 2C8 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction