missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (6402), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10564S
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TNF-alpha | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 6402 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| E. coli-derived recombinant human TNF-alpha , Gly57-Leu233, Accession # P01375 | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction