missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TNF-alpha Antibody (adalimumab), Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 893.00
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | adalimumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044197
|
Novus Biologicals™
NBP3-28186-100ug |
100 μg |
€ 349.00 € 329.70 / 100µg Save € 19.30 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046670
|
Novus Biologicals™
NBP3-28186-1mg |
1 mg | |||||||
|
30047068
|
Novus Biologicals™
NBP3-28186-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| TNF-alpha | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| TNFSF2/ TNFa | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| adalimumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P01375 | |
| 7124 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title