missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (B-F7), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14590MFV610
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for ELISA, Enzyme-linked Immunosorbent Spot
Specifications
| TNF-alpha | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, ELISpot | |
| B-F7 | |
| ELISA, Enzyme-linked Immunosorbent Spot | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Recombinant human TNF-alpha | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction