missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (Hu7) [DyLight 405], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals FAB9677E
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Neutralization, Flow Cytometry (Intracellular Staining)
Specifications
| TNF-alpha | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Neutralization, Flow Cytometry (Intracellular Staining) | |
| Hu7 | |
| Neutralization, Intracellular Staining by Flow Cytometry | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Human TNF-alpha | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction