missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNF beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585887
This item is not returnable.
View return policy
Description
TNF beta Polyclonal Antibody for Western Blot, ICC/IF
TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
Specifications
| TNF beta | |
| Polyclonal | |
| Unconjugated | |
| LTA | |
| DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25008, 4049 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 25% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P01374, Q06332 | |
| LTA | |
| Synthetic peptide within Human LT-alpha(C-term) aa 156-205. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction