missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Antibody (22221) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB226JF585
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal antibody specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Flow Cytometry,Neutralization,Immunocytochemistry
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| E. coli-derived recombinant human TNF RII, Leu23-Asp257, Accession # P20333 | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
| 7133 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Neutralization, Immunocytochemistry | |
| 22221 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction