missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Antibody (TR7554), Alexa Fluor™ 532, Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals FAB426X
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal antibody specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), CyTOF
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| TR7554 | |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| E. coli-derived recombinant mouse TNF RII/TNFRSF1B, Extracellular domain | |
| 0.5 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
| 7133 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction