missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUOX Monoclonal Antibody (OTI5A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805951S
This item is not returnable.
View return policy
Description
SUOX Monoclonal antibody specifically detects SUOX in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SUOX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07197 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4741 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A2 | |
| Unconjugated | |
| NEFM | |
| SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
| Human recombinant protein fragment corresponding to amino acids 413-773 of human NEFM produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction