missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNFAIP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15701151 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15701151 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15701151 Supplier Invitrogen™ Supplier No. PA553202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EARIYEETLN VLLYETPRVP DNSLLEATSR SRSQASPSED EETFELRDRV RRIHVKRYST YDDRQLGHQS THRD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNFAIP1
Gene Accession No. Q13829
Gene Alias B12; B61; BACURD2; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2; BTB/POZ domain-containing protein TNFAIP1; BTBD34; EDP1; Edp-1; hBACURD2; Protein B12; TNF alpha induced protein 1; Tnfaip1; Tnfip1; tumor necrosis factor induced protein 1; tumor necrosis factor, alpha induced protein 1; tumor necrosis factor, alpha-induced protein 1 (endothelial); Tumor necrosis factor, alpha-induced protein 1, endothelial; Tumor necrosis factor-induced protein 1
Gene Symbols TNFAIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNFAIP1. Recombinant protein control fragment (Product #RP-90724).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.