missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP8 Antibody (SR1270), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22595-50ul
This item is not returnable.
View return policy
Description
TNFAIP8 Monoclonal antibody specifically detects TNFAIP8 in Human samples. It is validated for Western Blot
Specifications
| TNFAIP8 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1270 | |
| Western Blot 1:500-1:2000 | |
| GG2-1, Head and neck tumor and metastasis-related protein, MDC-3.13TNF-induced protein GG2-1, NDED, NF-kappa-B-inducible DED-containing protein, SCC-S2SCCS2, TNF alpha-induced protein 8, tumor necrosis factor alpha-induced protein 8, tumor necrosis factor, alpha-induced protein 8 | |
| A synthesized peptide derived from human TNFAIP8 (Uniprot #: O95379) | |
| 50 μg | |
| Apoptosis | |
| 25816 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction