missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNFAIP8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA598204

Product Code. 16521032

  • € 473.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TNFAIP8 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TNFalpha-IP 8 (tumor necrosis factor, alpha-induced protein 8), also known as NFkappa B-inducible DED-containing protein (NDED), SCC-S2 or TNF-induced protein GG2-1, is a 198 amino acid cytoplasmic protein induced by NFkappa B and TNF. The induction of TNFalpha-IP 8 by TNF is dependent on the activation of NFkappa B. TNFalpha-IP 8 negatively mediates apoptosis and may also play a role in tumor progression. TNFalpha-IP 8 specifically inhibits caspase-8 activity, which results in the inhibition of BID cleavage and caspase-3 activation during TNF-mediated apoptosis. TNFalpha-IP 8 is expressed at high levels in thymus, bone marrow, lymph node, spleen, thyroid, placenta and various tumor tissues, as well as fetal lung, liver and kidney. TNFalpha-IP 8 is present as three isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNFAIP8
Polyclonal
Unconjugated
Tnfaip8
AA987150; E130304C20Rik; ENSMUSG00000073567; GG2-1; Gm10539; head and neck tumor and metastasis-related protein; MDC3.13; MDC-3.13; Nded; NF-kappa-B-inducible DED-containing protein; SCC S2; SCCS2; SCC-S2; Ssc-2; TNF alpha induced protein 8; TNF alpha-induced protein 8; Tnfaip8; TNFalpha IP8; TNFalpha-IP 8; TNFalphaIP8; TNF-induced protein; TNF-induced protein GG2-1; Tumor necrosis factor alpha-induced protein 8; tumor necrosis factor, alpha induced protein 8; tumor necrosis factor, alpha-induced protein 8
Rabbit
Antigen affinity chromatography
RUO
25816
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95379
Tnfaip8
Recombinant Human Tumor necrosis factor alpha-induced protein 8 protein (1-198AA).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.