missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP8L2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079626-B01P
This item is not returnable.
View return policy
Description
TNFAIP8L2 Polyclonal antibody specifically detects TNFAIP8L2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TNFAIP8L2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| FLJ23467, Inflammation factor protein 20, TIPE2, TIPE2inflammation factor 20, TNF alpha-induced protein 8-like protein 2, TNFAIP8-like protein 2, tumor necrosis factor alpha-induced protein 8-like protein 2, tumor necrosis factor, alpha-induced protein 8-like 2, tumor necrosis factor, alpha-induced protein 8-like protein 2 | |
| TNFAIP8L2 (AAH63014, 1 a.a. - 184 a.a.) full-length human protein. MESFSSKSLALQAEKKLLSKMAGRSVAHLFIDETSSEVLDELYRVSKEYTHSRPQAQRVIKDLIKVAIKVAVLHRNGSFGPSELALATRFRQKLRQGAMTALSFGEVDFTFEAAVLAGLLTECRDVLLELVEHHLTPKSHGRIRHVFDHFSDPGLLTALYGPDFTQHLGKICDGLRKLLDEGKL | |
| 0.05 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAH63014 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 79626 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction