Learn More
PeproTech TNFR2 (soluble) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Description
AA Sequence of recombinant protein: MAPEPGSTCR LREYYDQTAQ MCCSKCSPGQ HAKVFCTKTS DTVCDSCEDS TYTQLWNWVP ECLSCGSRCS SDQVETQACT REQNRICTCR PGWYCALSKQ EGCRLCAPLR KCRPGFGVAR PGTETSDVVC KPCAPGTFSN TTSSTDICRP HQICNVVAIP GNASMDAVCT STSP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human sTNF Receptor Type II. Anti-Human sTNF Receptor Type II-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hsTNF Receptor II by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human sTNF Receptor II (500-P168) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hsTNF Receptor II. Western Blot: To detect hsTNF Receptor II by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hsTNF Receptor II is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P168BT-1MG will be provided as 2 x 500 μg
Specifications
Specifications
| Antigen | TNFR2 (soluble) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with no preservative |
| Gene | Tnfrsf1b |
| Gene Accession No. | P20333 |
| Gene Alias | CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2 |
| Gene Symbols | Tnfrsf1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.