missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF10A Monoclonal Antibody (OTI11B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807650
This item is not returnable.
View return policy
Description
TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).Specifications
| TNFRSF10A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00220 | |
| TNFRSF10A | |
| Human recombit protein fragment corresponding to amino acids 263-468 of human TNFRSF10A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI11B12 | |
| Unconjugated | |
| TNFRSF10A | |
| APO2, CD261, DR4, TRAILR-1, TRAILR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8797 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction