missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF10A Monoclonal Antibody (OTI11B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807650S
This item is not returnable.
View return policy
Description
TNFRSF10A Monoclonal antibody specifically detects TNFRSF10A in Human samples. It is validated for Western BlotSpecifications
| TNFRSF10A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09919 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1440 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI11B12 | |
| Unconjugated | |
| Csf3 | |
| APO2; CD261; cytotoxic TRAIL receptor; death receptor 4; DR4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF receptor superfamily member 10a; TNF-related apoptosis-inducing ligand receptor 1; TNFRSF10A; TRAIL R1; TRAIL receptor 1; TRAILR1; TRAIL-R1; TRAILR-1; tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a | |
| Human recombinant protein fragment corresponding to amino acids 30-200 of human CSF3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction