missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF10A, Mouse anti-Human, Clone: TR1.02, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16095.100ug
This item is not returnable.
View return policy
Specifications
| TNFRSF10A | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| APO2/CD261/DR4/MGC9365/TRAILR-1/TRAILR1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| TR1.02 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| tumor necrosis factor receptor superfamily, member 10a | |
| TNFRSF10A | |
| Recombinant protein corresponding to human TNFRSF10A (DR4). | |
| RUO | |
| 8797 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction