missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR4 Mouse anti-Human, FITC, Clone: DR-4-02, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15746
This item is not returnable.
View return policy
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel member in the TNF family was recently identified and designated TRAIL (for TNF-related apoptosis-inducing ligand) and Apo-2L (for Apo-2 ligand). TRAIL is a type II membrane protein and expressed in a variety of human tissues. Two novel death domain containing receptors DR4 and DR5 have been identified as the receptor for TRAIL. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-βB activation in many tissues and cells.
Specifications
| DR4 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| O00220 | |
| TNFRSF10A | |
| Human CD261 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| DR-4-02 | |
| FITC | |
| TNFRSF10A | |
| APO2, CD261, DR4, MGC9365, TRAILR-1, TRAILR1 | |
| Mouse | |
| Purified | |
| RUO | |
| 8797 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction