missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF10B, Mouse anti-Human, Clone: TR2.21, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16096.100ug
This item is not returnable.
View return policy
Specifications
| TNFRSF10B | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| CD262/DR5/KILLER/KILLER/DR5/TRAIL-R2/TRAILR2/TRICK2/TRICK2A/TRICK2B/TRICKB/ZTNFR9 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| TR2.21 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| tumor necrosis factor receptor superfamily, member 10b | |
| TNFRSF10B | |
| Recombinant protein corresponding to human TNFRSF10B (DR5). | |
| RUO | |
| 8795 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction