missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFRSF11B Monoclonal Antibody (98A1071)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515960
This item is not returnable.
View return policy
Description
Suggested positive control: Daudi. Immunogen sequence: TQETFPPKYL HYDEETSH.
The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
Specifications
| TNFRSF11B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O00300, O08712, O08727 | |
| TNFRSF11B | |
| A peptide corresponding to the amino acids 20-37 of human Osteoprotegerin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 98A1071 | |
| Unconjugated | |
| TNFRSF11B | |
| LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin) | |
| Mouse | |
| Protein G | |
| RUO | |
| 100348133, 18383, 25341, 4982 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction