Learn More
TNFRSF14 Monoclonal Antibody (OTI7D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808066
Encompass_Preferred
 View more versions of this product
Product Code. 17578993
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
Specifications
Specifications
TNFRSF14
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q92956
TNFRSF14
Human recombit protein fragment corresponding to amino acids 39-283 of human TNFRSF14 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI7D7
Unconjugated
TNFRSF14
ATAR, CD270, HVEA, HVEM, LIGHTR, TR2
Mouse
Affinity Chromatography
RUO
8764
-20° C, Avoid Freeze/Thaw Cycles