Learn More
TNFRSF14 Mouse anti-Human, Clone: OTI1F12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808083
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.Specifications
| TNFRSF14 | |
| Monoclonal | |
| Unconjugated | |
| TNFRSF14 | |
| ATAR, CD270, HVEA, HVEM, LIGHTR, TR2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8764 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1F12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q92956 | |
| TNFRSF14 | |
| Human recombit protein fragment corresponding to amino acids 39-283 of human TNFRSF14 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.