Learn More
TNFRSF14 Monoclonal Antibody (OTI6E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813243S
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
TNFRSF14 Monoclonal antibody specifically detects TNFRSF14 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| TNFRSF14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92956 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8764 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI6E9 | |
| Unconjugated | |
| TNFRSF14 | |
| Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
| Full length human recombinant protein of human TNFRSF14 (NP_003811) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.