missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF18 Monoclonal Antibody (OTI4A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806458S
This item is not returnable.
View return policy
Description
TNFRSF18 Monoclonal antibody specifically detects TNFRSF18 in Human samples. It is validated for Western BlotSpecifications
| TNFRSF18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5U5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8784 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4A9 | |
| Unconjugated | |
| Tnfrsf18 | |
| Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
| Human recombinant protein fragment corresponding to amino acids 26-162 of human TNFRSF18 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction