missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF19 Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810325
This item is not returnable.
View return policy
Description
TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.Specifications
| TNFRSF19 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NS68 | |
| TNFRSF19 | |
| Human recombit protein fragment corresponding to amino acids 30-170 of human TNFRSF19 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B7 | |
| Unconjugated | |
| TNFRSF19 | |
| TAJ, TAJ-alpha, TRADE, TROY | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55504 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction