missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFRSF19L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584867
This item is not returnable.
View return policy
Description
Immunogen sequence: TSSMVSEVKT ITEAGPSWGD LPDSPQPGLP PEQQALLGSG GSRTKWLKPP AENKAEENRY VVRLSE Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 86%.
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is especially abundant in hematologic tissues. It has been shown to activate the NF-kappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1). This receptor is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported.
Specifications
| TNFRSF19L | |
| Polyclonal | |
| Unconjugated | |
| RELT | |
| E430021K24Rik; Receptor expressed in lymphoid tissues; Relt; RELT tumor necrosis factor receptor; Tnfrsf19l; TRLT; Tumor necrosis factor receptor superfamily member 19L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84957 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969Z4 | |
| RELT | |
| Recombinant protein corresponding to Human RELT. Recombinant protein control fragment (Product #RP-107822). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction