missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF8 Monoclonal Antibody (OTI13D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804026S
This item is not returnable.
View return policy
Description
TNFRSF8 Monoclonal antibody specifically detects TNFRSF8 in Human samples. It is validated for Flow CytometrySpecifications
| TNFRSF8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P28908 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 943 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry | |
| OTI13D8 | |
| Unconjugated | |
| TNFRSF8 | |
| Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30 | |
| Human recombinant protein fragment corresponding to amino acids 19-379 of human TNFRSF8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction