Learn More
TNFRSF8 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804027S
Encompass_Preferred
 View more versions of this product
Product Code. 17523353
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
TNFRSF8 Monoclonal antibody specifically detects TNFRSF8 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
TNFRSF8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P28908
Mouse
Affinity chromatography
RUO
943
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry
OTI1B2
Unconjugated
TNFRSF8
Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30
Human recombinant protein fragment corresponding to amino acids 19-379 of human TNFRSF8 produced in E.coli.
30 μL