Learn More
TNFRSF9 Mouse anti-Human, Clone: OTI5H5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF801748
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD137 is a member of the TNF-receptor superfamily and contributes to the clonal expansion, survival, and development of T cells. CD137 exists as both a monomer and a dimer on the surface of activated T cells. CD137 can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of CD137 is induced by lymphocyte activation. CD137 is also an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. CD137 exists as both a monomer and a dimer on the surface of activated T cells.Specifications
| TNFRSF9 | |
| Monoclonal | |
| Unconjugated | |
| TNFRSF9 | |
| 4-1BB, CD137, CDw137, ILA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3604 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5H5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q07011 | |
| TNFRSF9 | |
| Human recombit protein fragment corresponding to amino acids 24-186 of human TNFRSF9 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.