missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF15 Mouse anti-Human, Clone: VEGI/1283, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14901.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human TNFSF15.
- High specificity
- High reactivity
Specifications
| TNFSF15 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MGC129934/MGC129935/TL1/TL1A/VEGI/VEGI192A | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 9966 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| VEGI/1283 | |
| Mouse monoclonal antibody raised against full length recombinant human TNFSF15. | |
| In 10 mM PBS (0.05% BSA and 0.05% azide). | |
| TNFSF15 | |
| Recombinant protein corresponding to full-length human TNFSF15. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction