missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNIP3 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97865F
This item is not returnable.
View return policy
Description
TNIP3 Polyclonal specifically detects TNIP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TNIP3 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| A20, A20-binding inhibitor of NF-kappa-B activation 3, ABIN3, ABIN-3ABIN-3 beta, FLJ21162, LINDListeria induced, Listeria-induced gene protein, TNFAIP3 interacting protein 3, TNFAIP3-interacting protein 3, TNFAIP3-interacting protein 3 beta, TNIP3 beta | |
| Produced in rabbits immunized with E. coli-derived Human TNIP3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 79931 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction