missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNNC1 Monoclonal Antibody (OTI5H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804479
This item is not returnable.
View return policy
Description
Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.Specifications
| TNNC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P63316 | |
| TNNC1 | |
| Full length human recombit protein of human TNNC1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI5H3 | |
| Unconjugated | |
| TNNC1 | |
| CMD1Z, CMH13, TN-C, TNC, TNNC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7134 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction