missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNNC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596782
This item is not returnable.
View return policy
Description
Immunogen sequence: MTDQQAEARS YLSEEMIAEF KAAFDMFDAD GGGDISVKEL GTVMRMLGQT PTKEELDAII EEVDEDGSGT IDFEEFLVMM VRQMKEDAKG KSEEELAECF RIFDRNADGY IDPEELAEIF RASGEHVTDE EIESLMKDGD KNNDGRIDFD EFLKMMEGVQ; Positive Samples: U-937, HT-29, Mouse brain.
Troponin (Tn), a key protein complex in the regulation of striated muscle contraction, is composed of 3 subunits. The Tn-I subunit inhibits actomyosin ATPase, the Tn-T subunit binds tropomyosin and Tn-C, while the Tn-C subunit binds calcium and overcomes the inhibitory action of the troponin complex on actin filaments. The protein encoded by this gene is the Tn-C subunit.
Specifications
| TNNC2 | |
| Polyclonal | |
| Unconjugated | |
| TNNC2 | |
| skeletal troponin C; STNC; Tncs; Tnnc2; troponin C type 2 (fast); troponin C, fast skeletal; troponin C, skeletal muscle; troponin C2, fast; troponin C2, fast skeletal type | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21925, 296369, 7125 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02585, P20801 | |
| TNNC2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human TNNC2 (NP_0032701). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction