missing translation for 'onlineSavingsMsg'
Learn More

TNNI2 Monoclonal Antibody (OTI3A11), TrueMAB™, OriGene

Product Code. p-200016641
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17598703 30 μL 30µL
17578703 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17598703 Supplier Origene Technologies Supplier No. TA807853S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TNNI2 Monoclonal antibody specifically detects TNNI2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TNNI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pdcd1
Gene Accession No. Q02242, Q15116
Gene Alias AMCD2B; DA2B; FSSV; fsTnI; TnI; TNNI2; troponin 1, type 2; troponin I; troponin I fast twitch 2; troponin I type 2 (skeletal, fast); troponin I, fast skeletal muscle; Troponin I, fast-twitch isoform; troponin I, fast-twitch skeletal muscle isoform; troponin I, skeletal, fast; troponin I, skeletal, fast 2; troponin I2, fast skeletal type; Trp1
Host Species Mouse
Immunogen Full length human recombinant protein of human PDCD1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18566, 5133
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.