Learn More
TNNI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22130-1-AP-150UL
 View more versions of this product
Product Code. 16871665
  • € 410.00 / 150µL
Description
This gene encodes a fast-twitch skeletal muscle protein, a member of the troponin I gene family, and a component of the troponin complex including troponin T, troponin C and troponin I subunits. The troponin complex, along with tropomyosin, is responsible for the calcium-dependent regulation of striated muscle contraction. Mouse studies show that this component is also present in vascular smooth muscle and may play a role in regulation of smooth muscle function. In addition to muscle tissues, this protein is found in corneal epithelium, cartilage where it is an inhibitor of angiogenesis to inhibit tumor growth and metastasis, and mammary gland where it functions as a co-activator of estrogen receptor-related receptor alpha. This protein also suppresses tumor growth in human ovarian carcinoma. Mutations in this gene cause myopathy and distal arthrogryposis type 2B. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
TNNI2
Polyclonal
Unconjugated
TNNI2
AMCD2B, DA2B, FSSV, fsTnI, TNNI2, Troponin I fast skeletal muscle, Troponin I, fast-twitch isoform
Rabbit
Antigen Affinity Chromatography
RUO
21953, 29389, 7136
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13412, P27768, P48788
TNNI2
TNNI2 Fusion Protein Ag17750
150 μL
Primary
Human, Mouse, Rat