missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNNT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13423127
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13423127 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13423127 Supplier Invitrogen™ Supplier No. PA529003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, Porcine skeletal muscle. Predicted reactivity: Mouse (90%), Rat (90%), Pig (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is a subunit of troponin, which is a regulatory complex located on the thin filament of the sarcomere. This complex regulates striated muscle contraction in response to fluctuations in intracellular calcium concentration. This complex is composed of three subunits: troponin C, which binds calcium, troponin T, which binds tropomyosin, and troponin I, which is an inhibitory subunit. This protein is the slow skeletal troponin T subunit. Mutations in this gene cause nemaline myopathy type 5, also known as Amish nemaline myopathy, a neuromuscular disorder characterized by muscle weakness and rod-shaped, or nemaline, inclusions in skeletal muscle fibers which affects infants, resulting in death due to respiratory insufficiency, usually in the second year. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNNT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TNNT1
Gene Accession No. P13805, Q75ZZ6
Gene Alias ANM; AW146156; Fang2; MGC104241; NEM5; skeletal muscle slow-twitch TnT; slow skeletal muscle troponin T; ssTnT; sTnT; Tnnt1; TNT; TNTS; troponin T slow skeletal muscle type; troponin T slow type isoform sTnT1; troponin T slow type isoform sTnT2; troponin T type 1 (skeletal, slow); troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin T1, slow skeletal type; troponin-T1, skeletal, slow; zgc:193831; zgc:193865
Gene Symbols TNNT1
Host Species Rabbit
Immunogen Full length human Troponin T1 (slow) skeletal muscle Recombinant protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396579, 7138
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.