missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNNT2 Monoclonal Antibody (OTI5D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807904
This item is not returnable.
View return policy
Description
Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.Specifications
| TNNT2 | |
| Monoclonal | |
| Unconjugated | |
| TNNT2 | |
| CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7139 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5D9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P45379 | |
| TNNT2 | |
| Full length human recombit protein of human TNNT2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction