Learn More
Invitrogen™ TNNT3 Polyclonal Antibody, Biotin
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143042
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform 1, 2 and 4 (NP_006748.1; NP_001036246.1; NP_001036247.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
Specifications
| TNNT3 | |
| Polyclonal | |
| Biotin | |
| TNNT3 | |
| beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 21957, 24838, 7140 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P09739, P45378, Q9QZ47 | |
| TNNT3 | |
| Peptide with sequence CTTLRSRIDQAQKHSK (aa 240-254). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |