Learn More
Invitrogen™ TNNT3 Polyclonal Antibody, Biotin
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5143042
Product Code. 17848442
  • € 465.00 / 100µL
Description
Additional Information: This antibody is expected to recognize isoform 1, 2 and 4 (NP_006748.1; NP_001036246.1; NP_001036247.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNNT3
Polyclonal
Biotin
TNNT3
beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast
Goat
Ammonium sulfate precipitation
RUO
21957, 24838, 7140
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P09739, P45378, Q9QZ47
TNNT3
Peptide with sequence CTTLRSRIDQAQKHSK (aa 240-254).
100 μg
Primary
Human, Mouse, Rat