missing translation for 'onlineSavingsMsg'
Learn More

TNP, MedChemExpress

Product Code. 16460677
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460677

Brand: Medchem Express HY110079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 μM and 10.2 μM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes.

TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 μM and 10.2 μM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes.

Specifications

Chemical Name or Material TNP
Physical Form Solid
CAS 519178-28-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16F3N7O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 125 mg/mL (281.93 mM)
SMILES FC(C1=CC(CNC2=NC(NCC3=CC=C([N+]([O-])=O)C=C3)=C4N=CNC4=N2)=CC=C1)(F)F
Molecular Weight (g/mol) 443.38
Formula Weight 443.38
Percent Purity 98.79%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.