missing translation for 'onlineSavingsMsg'
Learn More

TNP1 Rabbit anti-Human, CoraLite 647, Polyclonal, Proteintech

Product Code. 17294846 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17294846 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17294846 Supplier Proteintech Supplier No. CL64717178100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Transition nuclear proteins (TNP1 and TNP2) are the major nuclear proteins that replace somatic histones during spermatogenesis. TNPs are required for normal chromatin condensation and functional sperm development, spermatogenesis was found to be compromised in both Tnp1 and Tnp2 null mice. TNP1, or TP1, localized in nucleus, is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Recently, TNP-1 was used as a germ cell marker in condensing spermatids.

Specifications

Antigen TNP1
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite 647
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 7141
Target Species Human
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.