missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNPO1 Monoclonal Antibody (D45)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA15885
This item is not returnable.
View return policy
Description
This antibody detects a band of approximately 90 kDa.
TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.
Specifications
| TNPO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8BFY9, Q92973 | |
| TNPO1 | |
| Recombinant his tagged transportin1 (Human) from SP2/0 cells. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rabbit, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| D45 | |
| Unconjugated | |
| TNPO1 | |
| AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455) | |
| Mouse | |
| Protein A | |
| RUO | |
| 100343169, 238799, 3842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction