missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNPO1 Monoclonal Antibody (D45)

Mouse Monoclonal Antibody
€ 522.00 - € 680.00
Specifications
| Antigen | TNPO1 |
|---|---|
| Clone | D45 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11594383
![]() |
Invitrogen™
MA15884 |
100 μL |
€ 680.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11504393
|
Invitrogen™
MA15885 |
50 μL |
€ 522.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects a band of approximately 90 kDa.
TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.Specifications
| TNPO1 | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.1% sodium azide | |
| Q8BFY9, Q92973 | |
| 100343169, 238799, 3842 | |
| Recombinant his tagged transportin1 (Human) from SP2/0 cells. | |
| Antibody |
| D45 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rabbit, Monkey | |
| TNPO1 | |
| AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455) | |
| TNPO1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title