missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNPO1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16500052
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16500052 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16500052 Supplier Invitrogen™ Supplier No. PA597214

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: LO2, U-87MG, DU145, HeLa, Mouse kidney, Rat kidney; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: CPQEVAPMLQ QFIRPWCTSL RNIRDNEEKD SAFRGICTMI SVNPSGVIQD FIFFCDAVAS WINPKDDLRD MFCKILHGFK NQVGDENWRR FSDQFPLPLK E.

TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNPO1
Gene Accession No. Q8BFY9, Q92973
Gene Alias AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455)
Gene Symbols TNPO1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 790-890 of human TNPO1 (NP_0022613).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238799, 309126, 3842
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.