missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNRC18B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569789
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rat: 100%; Zebrafish: 100%.
TNRC18P2 is a protein coding gene.
Specifications
| TNRC18B | |
| Polyclonal | |
| Unconjugated | |
| TNRC18P2 | |
| TNRC18P2 | |
| synthetic peptide directed towards the N terminal region of human TNRC18B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5-1.0 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27320 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction