missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNRC6B Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98079F
This item is not returnable.
View return policy
Description
TNRC6B Polyclonal specifically detects TNRC6B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TNRC6B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| KIAA1093trinucleotide repeat-containing gene 6B protein, trinucleotide repeat containing 6B | |
| Produced in rabbits immunized with E. coli-derived Human TNRC6B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 23112 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction