missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNRC6C Antibody (3G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057690-M08
This item is not returnable.
View return policy
Description
TNRC6C Monoclonal antibody specifically detects TNRC6C in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| TNRC6C | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| KIAA1582FLJ20015, trinucleotide repeat containing 6C, trinucleotide repeat-containing gene 6C protein | |
| TNRC6C (NP_061869, 181 a.a. ∽ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TDGPNNTNPMNSSPNPINAMQTNGLPNWGMAVGMGAIIPPHLQGLPGANGSSVSQVSGGSAEGISNSVWGLSPGNPATGNSNSGFSQGNGDTVNSALSAK | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| 3G11 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_061869 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 57690 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction