missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TO-PRO™3 Ready Flow™ Reagent

Product Code. 15721399
Change view
Click to view available options
Quantity:
120 assays
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15721399 120 assays 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15721399 Supplier Invitrogen™ Supplier No. R37170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TO-PRO™3 Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays.

  • A bright and easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays
  • Dye has excitation/emission maxima of 642/661 nm, similar to Alexa Fluor™ 647 or allophycocyanin (APC)
  • This direct-to-sample dead-cell stain emits red fluorescence when bound to DNA
  • Add 2 drops form the dropper bottle directly into 1 x 106 cells, incubate, and analyze
  • No need to dilute, weigh, or pipette
  • Stable at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Far-red
Content And Storage Includes 2 dropper bottles, store at room temperature.
Description TO-PRO™3 Ready Flow™ Reagent
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Reagent
Dye Type Cell-Impermeant
Emission 661
Excitation Wavelength Range 642 nm
Form Solution
Format Bottle(s)
Product Line Ready Flow, TO-PRO3
Quantity 120 assays
Solubility DMSO (Dimethylsulfoxide)
Sub Cellular Localization Nucleus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.