missing translation for 'onlineSavingsMsg'
Learn More

Toceranib Phosphate, TRC

Product Code. 30456366
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456366 50 mg -
30456001 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456366 Supplier Toronto Research Chemicals Supplier No. TRCT52550350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874819-74-6
Molecular Formula C22H25FN4O2 . H3PO4
Molecular Weight (g/mol) 494.5
Synonym PHA 291639E, 5-[(Z)-(5-Fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-N-[2-(1-pyrrolidinyl)ethyl]-1H-pyrrole-3-carboxamide Phosphate
IUPAC Name 5-[(Z)-(5-fluoro-2-oxo-1H-indol-3-ylidene)methyl]-2,4-dimethyl-N-(2-pyrrolidin-1-ylethyl)-1H-pyrrole-3-carboxamide;phosphoric acid
SMILES Cc1[nH]c(\C=C\2/C(=O)Nc3ccc(F)cc23)c(C)c1C(=O)NCCN4CCCC4.OP(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H25FN4O2.H3O4P/c1-13-19(12-17-16-11-15(23)5-6-18(16)26-21(17)28)25-14(2)20(13)22(29)24-7-10-27-8-3-4-9-27;1-5(2,3)4/h5-6,11-12,25H,3-4,7-10H2,1-2H3,(H,24,29)(H,26,28);(H3,1,2,3,4)/b17-12-;
Formula Weight 494.173
Chemical Name or Material Toceranib Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.