Learn More
Tocopherols (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Tocopherols |
| CAS | 1406-66-2 |
| Molecular Formula | C29 H50 O2 |
| Quantity | 50 g |
| Synonym | AT 160, Controx VP, Copherol F 1300, Covi-Ox T 50, Decanox MTS 50, Dry E-Mix 20, E-Mix 35L, ’ E-Oil 800, E-Oil Super 60, Fortium MTD 10, Golden Tocopherol 70, Irganox E 217DF, Riken Dry E-Mix SP-T, Riken E Oil Super 80G, Riken E-Emulsion, Tocoblend L 50IP, Tocomix L 70, Tocopherol 80, Tocovet E 1000, Tokomikkusu 500 |
| InChI Formula | InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21?,22?,29-/m0/s1 |
| SMILES | CC(C)CCCC(C)CCCC(C)CCC[C@@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1 |
| IUPAC Name | (2S)-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol |
| Molecular Weight (g/mol) | 430.706 |
| Formula Weight | 430.381 |