missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Glucagon-like peptide 1 (1-37) (human, rat)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Glucagon-like peptide 1 (1-37) (human, rat) is a pancreatic hormone synthesized by post-translational processing of proglucagon. Unlike truncated forms of GLP-1, it has no effect on food intake in rats and does not enhance pancreatic insulin secretion. However it induces insulin expression in intestinal epithelial cells, which can restore glucose homeostasis when implanted into diabetic mice.
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction